Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-methoxy-4-pyridineboronic acid 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44877-0010 44877-2500
TS44877-0010EA 180 USD InStock
TS44877-0010 TS44877-2500
3-Chloro-2-methoxy-4-pyridineboronic acid 95%
3-Chloro-2-methoxy-4-pyridineboronic acid


Formula: C₆H₇BClNO₃
Melting Pt: 140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06798255
CAS Number: 957060-88-7
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 137 to 142 °C
HPLC ≥94 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty