Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Cineole 85%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

45606-0250 45606-1000
TS45606-0250EA 41 USD InStock
TS45606-0250 TS45606-1000
1,4-Cineole 85%
1,4-Cineole


Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 65 °C (21hPa)
Melting Pt: –46 °C
Density: 0.887 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00209502
CAS Number: 470-67-7
UN: 1993
ADR: 3 III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4560 (20 °C, 589 nm)
GC ≥80 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty