Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis,cis-1,3,5-Cyclohexanetriol dihydrate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

45895-0010 45895-0050
TS45895-0010EA 55 USD InStock
TS45895-0010 TS45895-0050
cis,cis-1,3,5-Cyclohexanetriol dihydrate 98%
cis,cis-1,3,5-Cyclohexanetriol dihydrate


Formula: C₆H₁₂O₃·2H₂O
MW: 168.19 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00150671
CAS Number: 60662-54-6
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Water 19 to 24 %
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty