Smart SPME Fibers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5610-5870 5610-5871 5610-5872 5610-5873 5610-5874 5610-5875 5610-5876 5610-5878 5610-5879
AG5610-5870PK 880 USD InStock
AG5610-5870 AG5610-5871 AG5610-5872 AG5610-5873 AG5610-5874 AG5610-5875 AG5610-5876 AG5610-5878 AG5610-5879
Smart SPME Fibers, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Smart SPME fibers for PAL3 instrumentation.
  • Smart technology enables critical parameter tracking for each unique SPME fiber
  • Automatic application of the correct parameters for the individual smart SPME fiber
  • Compatible with all PAL3 models and existing SPME tools
  • Color coded to indicate the fiber coating and film thickness
  • Preinstalled with holder for easy maintenance
  • Smart SPME fibers kits available for method development
Agilent offers smart SPME fibers for compatibility with all PAL3 models including PAL 3 Series II smart instrumentation. Each smart SPME fiber is equipped with a unique smart chip containing parameters, ranges, and usage history for the specific fiber. Agilent smart SPME fibers are color coded to easily identify chemistry and thickness. Whether you are a new SPME user or switching to smart technology, you can be confident that performance of Agilent smart SPME fibers remains of the highest quality.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty