Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzoylbenzoic acid 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10571-0010 10571-0250 10571-5000
TS10571-0010EA 238 USD InStock
TS10571-0010 TS10571-0250 TS10571-5000
2-Benzoylbenzoic acid 98+%
2-Benzoylbenzoic acid


Formula: C₁₄H₁₀O₃
MW: 226.23 g/mol
Boiling Pt: 436
Melting Pt: 128
MDL Number: MFCD00002472
CAS Number: 85-52-9
EINECS: 201-612-2
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 126 to 130 °C
Titration with NaOH ≥98.0 %
UV at 375 nm A: ≤0.15
Viscosity 0.4 to 0.5 (1 % at 37 °C)
Average molecular weight 250000
pH 4.5 to 7.0 (6 % in water)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty