Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzylaminoethanol 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10594-1000 10594-5000
TS10594-1000EA 34 USD InStock
TS10594-1000 TS10594-5000
2-Benzylaminoethanol 96%
2-Benzylaminoethanol


Formula: C₆H₅CH₂NHCH₂CH₂OH
Boiling Pt: 153…156 °C (12 mmHg)
Density: 1.06 g/cm³
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00002840
CAS Number: 104-63-2
EINECS: 203-221-2
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5420 to 1.5440 (20°C, 589 nm)
GC ≥95 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty