Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromoanisole 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

10663-1000 10663-5000
TS10663-1000EA 91 USD InStock
TS10663-1000 TS10663-5000
4-Bromoanisole 98%
4-Bromoanisole


Formula: C₇H₇BrO
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.49 g/cm³
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00000097
CAS Number: 104-92-7
EINECS: 203-252-1
Merck Index: 13,01412
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5630 to 1.5650 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty