Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromopentafluorobenzene 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10722-0050 10722-0250 10722-1000
TS10722-0050EA 42 USD InStock
TS10722-0050 TS10722-0250 TS10722-1000
Bromopentafluorobenzene 99%
Bromopentafluorobenzene


Formula: C₆BrF₅
MW: 246.96 g/mol
Boiling Pt: 137
Melting Pt: -31
Density: 1.981 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000287
CAS Number: 344-04-7
EINECS: 206-449-0
Order Now
Specification Test Results
Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
Refractive index 1.4485 to 1.4505 (20°C, 589 nm)
GC ≥98.5 %
Absorption Ratio P-15/P+15 1.21 to 1.77
BSC Certification Passes

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty