Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromothiophenol 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

10746-0050 10746-0250 10746-1000
TS10746-0050EA 45 USD InStock
TS10746-0050 TS10746-0250 TS10746-1000
4-Bromothiophenol 95%
4-Bromothiophenol


Formula: C₆H₅BrS
Boiling Pt: 239 °C (1013 hPa)
Melting Pt: 71…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00004845
CAS Number: 106-53-6
EINECS: 203-407-3
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 71 to 76 °C
GC ≥94 %
Specific gravity (20/20 °C) 0.819 to 0.824
Acid value ≤8 mg KOH/g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty