Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloroaniline 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

10857-0010 10857-0050 10857-2500
TS10857-0010EA 101 USD InStock
TS10857-0010 TS10857-0050 TS10857-2500
2-Chloroaniline 98+%
2-Chloroaniline


Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 208
Melting Pt: -3
Density: 1.21 g/cm³
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00007656
CAS Number: 95-51-2
EINECS: 202-426-4
UN: 2019
ADR: 6.1 II
Merck Index: 13,02136
Order Now
Specification Test Results
Appearance Clear light yellow to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5870 to 1.5890 (20°C, 589 nm)
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty