Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chlorobenzaldehyde 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10861-0250 10861-1000
TS10861-0250EA 61 USD InStock
TS10861-0250 TS10861-1000
3-Chlorobenzaldehyde 99%
3-Chlorobenzaldehyde


Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 214
Melting Pt: 17…18 °C
Density: 1.241 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00003350
CAS Number: 587-04-2
EINECS: 209-596-9
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5630 to 1.5650 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty