Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

10993-0010 10993-0050 10993-0250
TS10993-0010EA 41 USD InStock
TS10993-0010 TS10993-0050 TS10993-0250
4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride 97%
4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride


Formula: C₁₁H₁₃Cl₂N
Melting Pt: 199…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00012753
CAS Number: 51304-61-1
UN: 2811
ADR: 6.1,II
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 199 to 204 °C
Titration with HClO4 ≥96.0 %
Distillation range 170 to 195 °C
Aromatics ≤50 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty