Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxybenzonitrile 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11082-0100 11082-0500
TS11082-0100EA 95 USD InStock
TS11082-0100 TS11082-0500
3-Hydroxybenzonitrile 97%
3-Hydroxybenzonitrile


Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 265
Melting Pt: 75…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00002252
CAS Number: 873-62-1
EINECS: 212-845-4
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 75 to 80 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty