Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibenzylamine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11261-0010 11261-2500
TS11261-0010EA 467 USD InStock
TS11261-0010 TS11261-2500
Dibenzylamine 98%
Dibenzylamine


Formula: C₆H₅CH₂NHCH₂C₆H₅
MW: 197.28 g/mol
Boiling Pt: 300
Melting Pt: -26
Density: 1.02 g/cm³
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00004770
CAS Number: 103-49-1
EINECS: 203-117-7
UN: 2735
ADR: 8 III
Merck Index: 13,03035
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5740 to 1.5760 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty