Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Di-tert-butylpyrocatechol 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11297-0050 11297-0250
TS11297-0050EA 32 USD InStock
TS11297-0050 TS11297-0250
3,5-Di-tert-butylpyrocatechol 99%
3,5-Di-tert-butylpyrocatechol


Formula: C₁₄H₂₂O₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 97…100 °C
MDL Number: MFCD00008819
CAS Number: 1020-31-1
EINECS: 213-816-9
Order Now
Specification Test Results
Appearance White to beige Crystalline powder or needles
Infrared spectrum Conforms
Melting point 97 to 100 °C
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty