Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichlone 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11348-0250 11348-1000 11348-2500
TS11348-0250EA 28 USD InStock
TS11348-0250 TS11348-1000 TS11348-2500
Dichlone 98%
Dichlone


Formula: C₁₀H₄Cl₂O₂
Boiling Pt: 275 °C (2 mmHg)
Melting Pt: 194…197 °C
MDL Number: MFCD00001677
CAS Number: 117-80-6
EINECS: 204-210-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,03071
Order Now
Specification Test Results
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 194 to 197 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty