Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Diethylamino)benzaldehyde 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11400-1000 11400-5000
TS11400-1000EA 61 USD InStock
TS11400-1000 TS11400-5000
4-(Diethylamino)benzaldehyde 99%
4-(Diethylamino)benzaldehyde


Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Boiling Pt: 174 °C (7 mmHg)
Melting Pt: 38…41 °C
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00003382
CAS Number: 120-21-8
EINECS: 204-377-4
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Green to brown to black Coarse crystals or chunks or crystalline solid
Infrared spectrum Conforms
Melting point 37 to 41 °C
Titration with HClO4 ≥98.5 %
Fluorescence max 605 nm (in Methanol)
Optimum lasing 614 nm (in Methanol)
Suitability (for use as a laser dye) Passes test
Molar absorption coefficient ≥105000 l/(mol.cm) (at Lambda max)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty