Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoroaniline 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11928-0250 11928-1000
TS11928-0250EA 51 USD InStock
TS11928-0250 TS11928-1000
3-Fluoroaniline 98%
3-Fluoroaniline


Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 186
Melting Pt: -2
Density: 1.15 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00007758
CAS Number: 372-19-0
EINECS: 206-747-0
UN: 2941
ADR: 6.1 III
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5430 to 1.5450 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty