Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fluorobenzene 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11934-0025 11934-1000 11934-5000
TS11934-0025EA 513 USD InStock
TS11934-0025 TS11934-1000 TS11934-5000
Fluorobenzene 99%
Fluorobenzene


Formula: C₆H₅F
MW: 96.1 g/mol
Boiling Pt: 85.1
Melting Pt: -41
Density: 1.02 g/cm³
Flash Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00000280
CAS Number: 462-06-6
EINECS: 207-321-7
UN: 2387
ADR: 3 II
Merck Index: 13,04197
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4643 to 1.4663 (20°C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty