Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-3-fluorotoulene 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11940-0050 11940-0250
TS11940-0050EA 35 USD InStock
TS11940-0050 TS11940-0250
α-Bromo-3-fluorotoulene 95%
α-Bromo-3-fluorotoulene


Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 88
Density: 1.541 g/cm³
Flash Pt: 61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000340
CAS Number: 456-41-7
EINECS: 207-263-2
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5480 (20°C, 589 nm)
GC ≥94 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty