Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxybenzophenone 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12101-0250 12101-1000
TS12101-0250EA 44 USD InStock
TS12101-0250 TS12101-1000
4-Hydroxybenzophenone 98+%
4-Hydroxybenzophenone


Formula: C₁₃H₁₀O₂
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00002355
CAS Number: 1137-42-4
EINECS: 214-507-1
Order Now
Specification Test Results
Appearance White to beige to brown Powder
Infrared spectrum Conforms
Melting point 132 to 135 °C
GC ≥98.0 %
Sodium chloride (NaCl) ≤200 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤0.5 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty