Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxybenzyl alcohol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12103-0250 12103-1000 12103-5000
TS12103-0250EA 111 USD InStock
TS12103-0250 TS12103-1000 TS12103-5000
4-Hydroxybenzyl alcohol 97%
4-Hydroxybenzyl alcohol


Formula: C₇H₈O₂
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004658
CAS Number: 623-05-2
EINECS: 210-768-0
Order Now
Specification Test Results
Appearance Beige to pink Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
TMS 0.028 to 0.032 v/v%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty