Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-1,2-Pentanediol 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

12329-0500 12329-2500
TS12329-0500EA 109 USD InStock
TS12329-0500 TS12329-2500
(±)-1,2-Pentanediol 96%
(±)-1,2-Pentanediol


Formula: C₅H₁₂O₂
Boiling Pt: 206 °C (760 mmHg)
Density: 0.971 g/cm³
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00010736
CAS Number: 5343-92-0
EINECS: 226-285-3
Order Now
Specification Test Results
Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20°C, 589 nm)
GC ≥95.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty