Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Chlorophenyl)acetyl chloride 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

12345-0050 12345-0250 12345-1000
TS12345-0050EA 34 USD InStock
TS12345-0050 TS12345-0250 TS12345-1000
(4-Chlorophenyl)acetyl chloride 96%
(4-Chlorophenyl)acetyl chloride


Formula: (C₆H₄Cl)CH₂COCl
Boiling Pt: 85 °C (1 mmHg)
Density: 1.292 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00037111
CAS Number: 25026-34-0
EINECS: 246-571-1
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5490 to 1.5570 (20°C, 589 nm)
GC ≥95.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty