Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((Diphenylmethylene)amino)acetonitrile 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

12414-0050 12414-0250
TS12414-0050EA 65 USD InStock
TS12414-0050 TS12414-0250
2-((Diphenylmethylene)amino)acetonitrile 99%
2-((Diphenylmethylene)amino)acetonitrile


Formula: C₁₅H₁₂N₂
MW: 220.2700 g/mol
Melting Pt: 83…86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009970
CAS Number: 70591-20-7
Order Now
Specification Test Results
Appearance White to yellow to light grey Powder or crystals
Infrared spectrum Conforms
Melting point 83 to 86 °C
HPLC ≥98.5 %
Identification A Passes test
Identification B Passes test
Specific optical rotation +12.0° to +12.8° (20 °C, 589 nm) (c=20, H2O) (on dried substance)
Titration with NaOH 99.5 to 101.0 % (on dried substance)
Sulfates ≤150 ppm
Chlorides ≤100 ppm
Calcium (Ca) ≤200 ppm
Impurity ≤350 ppm (as anhydrous oxalic acid)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty