Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Naphthoic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12818-0250 12818-1000
TS12818-0250EA 35 USD InStock
TS12818-0250 TS12818-1000
1-Naphthoic acid 98%
1-Naphthoic acid


Formula: C₁₁H₈O₂
MW: 172.18 g/mol
Boiling Pt: 300
Melting Pt: 162
MDL Number: MFCD00004007
CAS Number: 86-55-5
EINECS: 201-681-9
Merck Index: 12,06468
Order Now
Specification Test Results
Appearance Light yellow to brown Powder
Infrared spectrum Conforms
Melting point 160 to 164 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty