Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dihydroxytoluene monohydrate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12955-0250 12955-1000 12955-5000
TS12955-0250EA 117 USD InStock
TS12955-0250 TS12955-1000 TS12955-5000
3,5-Dihydroxytoluene monohydrate 99%
3,5-Dihydroxytoluene monohydrate


Formula: C₇H₈O₂·1H₂O
Boiling Pt: 290 °C (760 mmHg)
Melting Pt: 56…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00149092
CAS Number: 6153-39-5
EINECS: 207-984-2
Merck Index: 12,06995
Order Now
Specification Test Results
Appearance White to beige to pink-brown Powder or crystals
Infrared spectrum Conforms
Melting point 56 to 61 °C
Water 10 to 14 % (K.F.)
Solubility (5 % in water) Clear colorless to light brown
HPLC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty