Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

o-Phenylenediacetonitrile 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

13052-0100 13052-0500
TS13052-0100EA 24 USD InStock
TS13052-0100 TS13052-0500
o-Phenylenediacetonitrile 99+%
o-Phenylenediacetonitrile


Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00001905
CAS Number: 613-73-0
EINECS: 210-351-3
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to yellow to beige Crystalline powder or crystals (May contain yellow to orange chunks)
Infrared spectrum Conforms
Melting point 57 to 61 °C
GC ≥99.0 %
Residue after evaporation ≤0.001 %
Filtered to 0.2 micrometer Filtered

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty