Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Phenylpropanal 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

13086-0250 13086-1000 13086-5000
TS13086-0250EA 22 USD InStock
TS13086-0250 TS13086-1000 TS13086-5000
(±)-2-Phenylpropanal 98%
(±)-2-Phenylpropanal


Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 202…205 °C (760 mmHg)
Melting Pt: –60 °C
Density: 1.011 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00006973
CAS Number: 93-53-8
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5160 to 1.5240 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty