Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Salicylohydroxamic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

13262-0050 13262-0250
TS13262-0050EA 33 USD InStock
TS13262-0050 TS13262-0250
Salicylohydroxamic acid 99%
Salicylohydroxamic acid


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 177…185 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002110
CAS Number: 89-73-6
EINECS: 201-934-3
Order Now
Specification Test Results
Appearance Pink-beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Elemental Analysis 8.67 to 9.11 % N
Specific optical rotation +6.5° to +9.5° (20°C, 589 nm) c=5 in 1N HCl

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty