Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Diphenyl-2-thiourea 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

13870-0010 13870-0025 13870-1000
TS13870-0010EA 89 USD InStock
TS13870-0010 TS13870-0025 TS13870-1000
1,3-Diphenyl-2-thiourea 98%
1,3-Diphenyl-2-thiourea


Formula: C₁₃H₁₂N₂S
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00004921
CAS Number: 102-08-9
EINECS: 203-004-2
UN: 3077
ADR: 9,III
Merck Index: 13,03366
Order Now
Specification Test Results
Appearance White to cream to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 148 to 154 °C
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty