Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiophenol 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

13887-0010 13887-0050
TS13887-0010EA 186 USD InStock
TS13887-0010 TS13887-0050
Thiophenol 99%
Thiophenol


Formula: C₆H₆S
MW: 110.18 g/mol
Boiling Pt: 167…169 °C (760 mmHg)
Melting Pt: –15 °C
Density: 1.078 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00004826
CAS Number: 108-98-5
EINECS: 203-635-3
UN: 2337
ADR: 6.1
Merck Index: 12,09492
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5880 to 1.5900 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty