Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

p-Toluic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

13906-0010 13906-1000 13906-2500
TS13906-0010EA 137 USD InStock
TS13906-0010 TS13906-1000 TS13906-2500
p-Toluic acid 98%
p-Toluic acid


Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 275
Melting Pt: 179…182 °C
Flash Pt: 180
Storage Temperature: Ambient
MDL Number: MFCD00002565
CAS Number: 99-94-5
EINECS: 202-803-3
Merck Index: 12,09673
Order Now
Specification Test Results
Appearance White to cream to light yellow Powder
Infrared spectrum Conforms
Melting point 179 to 182 °C
Titration with NaOH ≥97.5 %
Elemental Analysis 41.2 to 46.4 % (O)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty