Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dichlorobenzoyl chloride 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15023-0050 15023-0250 15023-1000
TS15023-0050EA 26 USD InStock
TS15023-0050 TS15023-0250 TS15023-1000
3,4-Dichlorobenzoyl chloride 97%
3,4-Dichlorobenzoyl chloride


Formula: C₇H₃Cl₃O
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 30…33 °C
Flash Pt: 142 °C
Storage Temperature: Ambient
MDL Number: MFCD00000672
CAS Number: 3024-72-4
EINECS: 221-177-2
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White to yellow Low melting crystalline solid
Infrared spectrum Conforms
GC ≥96.0 %
Titration RCOCl ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty