Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitro-o-toluidine 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15202-0250 15202-1000
TS15202-0250EA 53 USD InStock
TS15202-0250 TS15202-1000
3-Nitro-o-toluidine 97%
3-Nitro-o-toluidine


Formula: C₇H₈N₂O₂
Boiling Pt: 305 °C (760 mmHg)
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007731
CAS Number: 603-83-8
EINECS: 210-059-6
UN: 2660
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Yellow to yellow-green Crystalline powder
Infrared spectrum Conforms
Melting point 87 to 91 °C
HPLC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty