Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminothiophenol 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15390-1000 15390-5000
TS15390-1000EA 98 USD InStock
TS15390-1000 TS15390-5000
2-Aminothiophenol 98%
2-Aminothiophenol


Formula: C₆H₇NS
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 20 °C
Density: 1.16 g/cm³
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007702
CAS Number: 137-07-5
EINECS: 205-277-3
UN: 3267
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.6410 to 1.6450 (20°C, 589 nm)
Titration Argentometric 97.5 to 102.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty