Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloro(diphenyl)methane 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15466-0050 15466-0500 15466-2500
TS15466-0050EA 32 USD InStock
TS15466-0050 TS15466-0500 TS15466-2500
Chloro(diphenyl)methane 98%
Chloro(diphenyl)methane


Formula: (C₆H₅)₂CHCl
MW: 202.68 g/mol
Boiling Pt: 315
Melting Pt: 17
Density: 1.14 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000855
CAS Number: 90-99-3
EINECS: 202-031-7
UN: 3265
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5940 to 1.5970 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty