Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,4-Dimethoxyphenyl)acetonitrile 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15526-0250 15526-1000 15526-5000
TS15526-0250EA 27 USD InStock
TS15526-0250 TS15526-1000 TS15526-5000
(3,4-Dimethoxyphenyl)acetonitrile 99+%
(3,4-Dimethoxyphenyl)acetonitrile


Formula: C₁₀H₁₁NO₂
Boiling Pt: 171…178 °C (10 mmHg)
Melting Pt: 62…65 °C
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00001911
CAS Number: 93-17-4
EINECS: 202-225-1
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to off-white Crystals or crystalline needles
Infrared spectrum Conforms
Melting point 62 to 65 °C
Loss on drying ≤0.5 % (vacuum)
Sulfated ash ≤0.1 %
GC ≥99.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty