Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluorobenzoic acid 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

15616-0250 15616-1000 15616-5000
TS15616-0250EA 34 USD InStock
TS15616-0250 TS15616-1000 TS15616-5000
4-Fluorobenzoic acid 99%
4-Fluorobenzoic acid


Formula: C₇H₅FO₂
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 184…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00002530
CAS Number: 456-22-4
EINECS: 207-259-0
Merck Index: 13,04198
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 184 to 187 °C
GC ≥98.5 %
Titration with NaOH ≥98.5 %
Residue after evaporation ≤0.001 %
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Potassium (K) ≤0.5 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty