Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tribenzylamine 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

15647-0010 15647-1000 15647-2500
TS15647-0010EA 283 USD InStock
TS15647-0010 TS15647-1000 TS15647-2500
Tribenzylamine 99+%
Tribenzylamine


Formula: N(CH₂C₆H₅)₃
Boiling Pt: 380…390 °C (760 mmHg)
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00004773
CAS Number: 620-40-6
EINECS: 210-638-3
Order Now
Specification Test Results
Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 90 to 94 °C
Water ≤0.5 % (K.F.)
GC ≥99.0 %
Titration with HClO4 ≥99.0 % (On anhydrous substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty