Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Isopropylphenol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15668-0050 15668-5000
TS15668-0050EA 25 USD InStock
TS15668-0050 TS15668-5000
2-Isopropylphenol 97%
2-Isopropylphenol


Formula: C₉H₁₂O
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 15…16 °C
Density: 1.01 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00002224
CAS Number: 88-69-7
EINECS: 201-852-8
UN: 2922
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5290 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty