Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trimethylbenzyl chloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15978-0050 15978-0250
TS15978-0050EA 33 USD InStock
TS15978-0050 TS15978-0250
2,4,6-Trimethylbenzyl chloride 98%
2,4,6-Trimethylbenzyl chloride


Formula: C₁₀H₁₃Cl
Boiling Pt: 114…115 °C (10 mmHg)
Melting Pt: 34…38 °C
Flash Pt: 106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000901
CAS Number: 1585-16-6
EINECS: 216-440-3
UN: 3261
ADR: 8,II
Order Now
Specification Test Results
Appearance White Fused solid or crystals and/or chunks
Infrared spectrum Conforms
Melting point 37 to 42 °C
GC ≥97.5 %
Residue after evaporation ≤0.0005 %
Acidity (CH3COOH) ≤0.002 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty