Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,3-Trimethoxybenzene 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

16021-0250 16021-1000
TS16021-0250EA 42 USD InStock
TS16021-0250 TS16021-1000
1,2,3-Trimethoxybenzene 98%
1,2,3-Trimethoxybenzene


Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 241
Melting Pt: 42…45 °C
Density: 1.112 g/cm³
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00008358
CAS Number: 634-36-6
EINECS: 211-207-2
Order Now
Specification Test Results
Appearance White to beige to light brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 42 to 47 °C
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty