Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethylbenzoic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

16033-0100 16033-0500
TS16033-0100EA 67 USD InStock
TS16033-0100 TS16033-0500
2,5-Dimethylbenzoic acid 98%
2,5-Dimethylbenzoic acid


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 268
Melting Pt: 131…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00002482
CAS Number: 610-72-0
EINECS: 210-235-2
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 131 to 135 °C
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty