Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxyphenol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16106-0050 16106-0250 16106-1000 16106-5000
TS16106-0050EA 26 USD InStock
TS16106-0050 TS16106-0250 TS16106-1000 TS16106-5000
3-Methoxyphenol 97%
3-Methoxyphenol


Formula: C₇H₈O₂
Boiling Pt: 244 °C (760 mmHg)
Melting Pt: –17 °C
Density: 1.145 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002267
CAS Number: 150-19-6
EINECS: 205-754-6
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear pink-red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5510 to 1.5530 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty