Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-p-toluonitrile 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16276-0100 16276-0500
TS16276-0100EA 88 USD InStock
TS16276-0100 TS16276-0500
α-Bromo-p-toluonitrile 98%
α-Bromo-p-toluonitrile


Formula: C₈H₆BrN
Boiling Pt: 135…137 °C (0.6 mmHg)
Melting Pt: 113…117 °C
MDL Number: MFCD00001829
CAS Number: 17201-43-3
EINECS: 241-246-0
UN: 1759
ADR: 8,II
Order Now
Specification Test Results
Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 113 to 118 °C
GC ≥97.5 %
Iron (Fe) ≤0.1 %
Phosphorus (P) ≤0.04 %
Potassium (K) ≤0.4 %
Silicon (Si) ≤0.06 %
Sodium (Na) ≤0.3 %
Sulfur (S) ≤0.07 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty