Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Hydroxy-m-toluic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

16386-0100 16386-0500
TS16386-0100EA 82 USD InStock
TS16386-0100 TS16386-0500
6-Hydroxy-m-toluic acid 98%
6-Hydroxy-m-toluic acid


Formula: C₈H₈O₃
Melting Pt: 151…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00002461
CAS Number: 89-56-5
EINECS: 201-918-6
Merck Index: 12,02650
Order Now
Specification Test Results
Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 151 to 155 °C
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty