Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxybenzylamine 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

17054-0100 17054-0500
TS17054-0100EA 47 USD InStock
TS17054-0100 TS17054-0500
2-Methoxybenzylamine 98%
2-Methoxybenzylamine


Formula: C₈H₁₁NO
Boiling Pt: 231…233 °C (760 mmHg)
Density: 1.05 g/cm³
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008110
CAS Number: 6850-57-3
EINECS: 229-941-7
UN: 2735
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5475 to 1.5495 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty