Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Salicyl alcohol 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

17333-0010 17333-0250 17333-1000
TS17333-0010EA 967 USD InStock
TS17333-0010 TS17333-0250 TS17333-1000
Salicyl alcohol 97%
Salicyl alcohol


Formula: C₇H₈O₂
Melting Pt: 83…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00004617
CAS Number: 90-01-7
EINECS: 201-960-5
Merck Index: 12,08477
Order Now
Specification Test Results
Appearance Light beige Powder or flakes
Infrared spectrum Conforms
Melting point 83 to 87 °C
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty