Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-nitrobenzene 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

17336-0100 17336-0250 17336-1000
TS17336-0100EA 37 USD InStock
TS17336-0100 TS17336-0250 TS17336-1000
1-Bromo-4-nitrobenzene 98%
1-Bromo-4-nitrobenzene


Formula: C₆H₄BrNO₂
Boiling Pt: 255 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00007280
CAS Number: 586-78-7
EINECS: 209-583-8
UN: 3459
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty